siRNA Details
| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007750.1 | NP_001007751.1 | 5 | 795 | ||
| XM_006241228.2 | XP_006241290.1 | 5 | 790 | |||
| XM_006241229.2 | XP_006241291.1 | 5 | 790 | |||
| XM_006241230.2 | XP_006241292.1 | 5 | 790 | |||
| XM_006241231.2 | XP_006241293.1 | 5 | 790 | |||
| GenBank mRNA | BC082074.1 | - | 5 | 795 | 7133149 | |

Chpt1
Genomic Map