siRNA Details
| Gene Symbol: Os9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007265.1 | NP_001007266.1 | 3, 4 | 432 | ||
| XM_006241461.1 | XP_006241523.1 | 3, 4 | 453 | |||
| GenBank mRNA | BC085907.1 | - | 3, 4 | 432 | 7096059 | |
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Mouse: NM_001171026, NM_177614, BC006844, BC031768, BU703218, and others.
					            	This siRNA also targets the following transcripts in Mouse: NM_001171026, NM_177614, BC006844, BC031768, BU703218, and others.Gene Ontology Categories:
 Back To Top
 Back To Top
 Os9
Os9 Genomic Map
Genomic Map Back To Top
 Back To Top