siRNA Details
| Gene Symbol: Stat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044250.1 | NP_001037715.1 | 18 | 2125 | ||
| GenBank mRNA | DQ462201.1 | - | 18 | 2125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stat6| Gene Symbol: Stat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044250.1 | NP_001037715.1 | 18 | 2125 | ||
| GenBank mRNA | DQ462201.1 | - | 18 | 2125 | ||