siRNA Details
| Gene Symbol: MGC94207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007751.1 | NP_001007752.1 | 3 | 549 | ||
| GenBank mRNA | BC081992.1 | - | 3 | 549 | 7130152 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC94207| Gene Symbol: MGC94207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007751.1 | NP_001007752.1 | 3 | 549 | ||
| GenBank mRNA | BC081992.1 | - | 3 | 549 | 7130152 | |