siRNA Details
| Gene Symbol: Sgsm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198787.2 | NP_942082.2 | 16 | 1803 | ||
| XM_006242096.2 | XP_006242158.1 | 16 | 1829 | |||
| XM_006242098.2 | XP_006242160.1 | 16 | 1834 | |||
| XM_006242099.2 | XP_006242161.1 | 16 | 1834 | |||
| XM_008765761.1 | XP_008763983.1 | 16 | 1834 | |||
| XM_008765762.1 | XP_008763984.1 | 16 | 3535 | |||
| XM_008765763.1 | XP_008763985.1 | 16 | 1829 | |||
| GenBank mRNA | AB091009.2 | - | 14 | 1651 | ||
| BC062060.1 | - | 16 | 1770 | 5599107 | ||
| FQ212274.1 | - | 16 | 1801 | |||

Sgsm3
Genomic Map