siRNA Details
| Gene Symbol: Sgsm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198787.2 | NP_942082.2 | 9, 10 | 1110 | ||
| XM_006242096.2 | XP_006242158.1 | 9, 10 | 1136 | |||
| XM_006242098.2 | XP_006242160.1 | 9, 10 | 1141 | |||
| XM_006242099.2 | XP_006242161.1 | 9, 10 | 1141 | |||
| XM_008765761.1 | XP_008763983.1 | 9, 10 | 1141 | |||
| XM_008765762.1 | XP_008763984.1 | 9, 10 | 2842 | |||
| XM_008765763.1 | XP_008763985.1 | 9, 10 | 1136 | |||
| GenBank mRNA | AB091009.2 | - | 7, 8 | 958 | ||
| BC062060.1 | - | 9, 10 | 1077 | 5599107 | ||
| FQ212274.1 | - | 9, 10 | 1108 | |||

Sgsm3
Genomic Map