siRNA Details
| Gene Symbol: Panx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199409.2 | NP_955441.2 | 2 | 944 | ||
| GenBank mRNA | AJ557016.1 | - | 2 | 944 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Panx2| Gene Symbol: Panx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199409.2 | NP_955441.2 | 2 | 944 | ||
| GenBank mRNA | AJ557016.1 | - | 2 | 944 | ||