siRNA Details
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108750.1 | NP_001102220.1 | 16 | 1411 | ||
GenBank mRNA | AM747284.1 | - | 16 | 1282 | ||
BC168840.1 | - | 16 | 1334 | 7125016 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108750.1 | NP_001102220.1 | 16 | 1411 | ||
GenBank mRNA | AM747284.1 | - | 16 | 1282 | ||
BC168840.1 | - | 16 | 1334 | 7125016 |