siRNA Details
| Gene Symbol: Zcrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034940.1 | NP_001030112.1 | 4 | 284 | ||
| GenBank mRNA | BC099747.1 | - | 4 | 284 | 5598773 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcrb1| Gene Symbol: Zcrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034940.1 | NP_001030112.1 | 4 | 284 | ||
| GenBank mRNA | BC099747.1 | - | 4 | 284 | 5598773 | |