siRNA Details
Gene Symbol: Ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014203.1 | NP_001014225.1 | 8 | 1437 | ||
GenBank mRNA | BC083683.1 | - | 8 | 1437 | 7135919 | |
FQ212708.1 | - | 6 | 1199 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014203.1 | NP_001014225.1 | 8 | 1437 | ||
GenBank mRNA | BC083683.1 | - | 8 | 1437 | 7135919 | |
FQ212708.1 | - | 6 | 1199 |