siRNA Details
| Gene Symbol: Ccdc65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014203.1 | NP_001014225.1 | 3 | 452 | ||
| XM_006257362.2 | XP_006257424.1 | 3 | 518 | |||
| GenBank mRNA | BC083683.1 | - | 3 | 452 | 7135919 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc65| Gene Symbol: Ccdc65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014203.1 | NP_001014225.1 | 3 | 452 | ||
| XM_006257362.2 | XP_006257424.1 | 3 | 518 | |||
| GenBank mRNA | BC083683.1 | - | 3 | 452 | 7135919 | |