siRNA Details
| Gene Symbol: Slc44a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134715.1 | NP_001128187.1 | 14, 15 | 1304 | ||
| XM_006242649.2 | XP_006242711.1 | 14, 15 | 2054 | |||
| XM_006242650.2 | XP_006242712.1 | 14, 15 | 1324 | |||
| XM_006242651.2 | XP_006242713.1 | 14, 15 | 1324 | |||
| GenBank mRNA | BC168183.1 | - | 14, 15 | 1304 | 7098013 | |
| FQ225791.1 | - | 14, 15 | 1325 | |||
| FQ231292.1 | - | 14, 15 | 1325 | |||

Slc44a2
Genomic Map