siRNA Details
| Gene Symbol: Carm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030041.3 | NP_001025212.1 | 5, 6 | 769 | ||
| NM_001034088.2 | NP_001029260.1 | 5, 6 | 769 | |||
| GenBank mRNA | AB201114.1 | - | 5, 6 | 769 | ||
| AB201115.1 | - | 5, 6 | 769 | |||
| AB201116.1 | - | 5, 6 | 769 | |||
| AB201117.1 | - | 5, 6 | 769 | |||
| KJ672502.1 | - | 5, 6 | 679 | |||

Carm1
Genomic Map