siRNA Details
| Gene Symbol: Swsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108755.1 | NP_001102225.1 | 2 | 811 | ||
| GenBank mRNA | BC168995.1 | - | 2 | 769 | 7193660 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Swsap1| Gene Symbol: Swsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108755.1 | NP_001102225.1 | 2 | 811 | ||
| GenBank mRNA | BC168995.1 | - | 2 | 769 | 7193660 | |