siRNA Details
Gene Symbol: Stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029924.1 | NP_001025095.1 | 6 | 545 | ||
GenBank mRNA | BC097373.1 | - | 6 | 545 | 7444462 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029924.1 | NP_001025095.1 | 6 | 545 | ||
GenBank mRNA | BC097373.1 | - | 6 | 545 | 7444462 |