siRNA Details
| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108770.1 | NP_001102240.1 | 8 | 1607 | ||
| XM_006243332.2 | XP_006243394.1 | 10 | 1720 | |||
| XM_006243337.2 | XP_006243399.1 | 9 | 1663 | |||
| XM_006243341.2 | XP_006243403.1 | 8 | 1636 | |||
| XM_008766315.1 | XP_008764537.1 | 10 | 1980 | |||
| XM_008766316.1 | XP_008764538.1 | 10 | 1833 | |||
| XM_008766317.1 | XP_008764539.1 | 9 | 1693 | |||
| XM_008766318.1 | XP_008764540.1 | 10 | 1720 | |||
| XM_008766319.1 | XP_008764541.1 | 8 | 1636 | |||
| XM_008766320.1 | XP_008764542.1 | 10 | 1720 | |||

Ccpg1
Genomic Map