siRNA Details
| Gene Symbol: Rsl24d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014212.1 | NP_001014234.1 | 5 | 441 | ||
| GenBank mRNA | BC062237.1 | - | 5 | 441 | 6919069 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsl24d1| Gene Symbol: Rsl24d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014212.1 | NP_001014234.1 | 5 | 441 | ||
| GenBank mRNA | BC062237.1 | - | 5 | 441 | 6919069 | |