siRNA Details
| Gene Symbol: Slc17a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009713.2 | NP_001009713.1 | 6 | 787 | ||
| GenBank mRNA | AY800277.1 | - | 6 | 786 | ||
| BC097482.1 | - | 6 | 787 | 7459136 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a5| Gene Symbol: Slc17a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009713.2 | NP_001009713.1 | 6 | 787 | ||
| GenBank mRNA | AY800277.1 | - | 6 | 786 | ||
| BC097482.1 | - | 6 | 787 | 7459136 | ||