siRNA Details
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047916.1 | NP_001041381.1 | 9 | 1072 | ||
XM_006243513.2 | XP_006243575.1 | 1810 | ||||
XM_006243514.1 | XP_006243576.1 | 1627 | ||||
XM_006243515.1 | XP_006243577.1 | 1765 | ||||
XM_006243516.1 | XP_006243578.1 | 1765 | ||||
XM_006243517.2 | XP_006243579.1 | 8 | 1037 | |||
XM_006243518.2 | XP_006243580.1 | 10 | 1261 | |||
XM_006243519.2 | XP_006243581.1 | 10 | 1261 | |||
GenBank mRNA | AY325202.1 | - | 9 | 1072 |
