siRNA Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108775.1 | NP_001102245.1 | 3, 4 | 219 | ||
| XM_006243669.2 | XP_006243731.1 | 4, 5 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab6b| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108775.1 | NP_001102245.1 | 3, 4 | 219 | ||
| XM_006243669.2 | XP_006243731.1 | 4, 5 | 523 | |||