siRNA Details
| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191883.1 | NP_001178812.1 | 12, 13 | 1133 | ||
| XM_006243802.2 | XP_006243864.1 | 13, 14 | 1629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcbp4| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191883.1 | NP_001178812.1 | 12, 13 | 1133 | ||
| XM_006243802.2 | XP_006243864.1 | 13, 14 | 1629 | |||