siRNA Details
| Gene Symbol: Zmynd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004284.1 | NP_001004284.1 | 9 | 1026 | ||
| GenBank mRNA | BC079255.1 | - | 9 | 1026 | 7133212 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmynd10| Gene Symbol: Zmynd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004284.1 | NP_001004284.1 | 9 | 1026 | ||
| GenBank mRNA | BC079255.1 | - | 9 | 1026 | 7133212 | |