siRNA Details
| Gene Symbol: Gnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108780.2 | NP_001102250.2 | 8 | 994 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnat1| Gene Symbol: Gnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108780.2 | NP_001102250.2 | 8 | 994 | ||