siRNA Details
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014214.1 | NP_001014236.1 | 8 | 1078 | ||
GenBank mRNA | BC082046.1 | - | 8 | 1078 | 7131735 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014214.1 | NP_001014236.1 | 8 | 1078 | ||
GenBank mRNA | BC082046.1 | - | 8 | 1078 | 7131735 |