siRNA Details
| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108782.2 | NP_001102252.1 | 5 | 366 | ||
| GenBank mRNA | BC166773.1 | - | 5 | 366 | 7934150 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elp6| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108782.2 | NP_001102252.1 | 5 | 366 | ||
| GenBank mRNA | BC166773.1 | - | 5 | 366 | 7934150 | |