siRNA Details
| Gene Symbol: Csrnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108786.1 | NP_001102256.1 | 5 | 1431 | ||
| XM_006244119.2 | XP_006244181.1 | 5 | 2098 | |||
| XM_006244120.2 | XP_006244182.1 | 5 | 1703 | |||
| XM_006244121.1 | XP_006244183.1 | 5 | 3854 | |||
| XM_006244122.2 | XP_006244184.1 | 5 | 1448 | |||
| XM_008766688.1 | XP_008764910.1 | 4 | 4771 | |||
| XM_008766689.1 | XP_008764911.1 | 5 | 3715 | |||

Csrnp1
Genomic Map