siRNA Details
| Gene Symbol: Wdr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199410.2 | NP_955442.1 | 6 | 713 | ||
| GenBank mRNA | BC063180.1 | - | 6 | 717 | 6920424 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr12| Gene Symbol: Wdr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199410.2 | NP_955442.1 | 6 | 713 | ||
| GenBank mRNA | BC063180.1 | - | 6 | 717 | 6920424 | |