siRNA Details
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177419.2 | NP_803154.1 | 8 | 936 | ||
GenBank mRNA | AB066103.1 | - | 6 | 693 | ||
BC061576.1 | - | 8 | 939 | 6887693 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177419.2 | NP_803154.1 | 8 | 936 | ||
GenBank mRNA | AB066103.1 | - | 6 | 693 | ||
BC061576.1 | - | 8 | 939 | 6887693 |