siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100760.1 | NP_001094230.1 | 6, 7 | 846 | ||
| XM_006245244.2 | XP_006245306.1 | 6, 7 | 868 | |||
| GenBank mRNA | BC104673.1 | - | 6, 7 | 656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam134a| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100760.1 | NP_001094230.1 | 6, 7 | 846 | ||
| XM_006245244.2 | XP_006245306.1 | 6, 7 | 868 | |||
| GenBank mRNA | BC104673.1 | - | 6, 7 | 656 | ||