siRNA Details
| Gene Symbol: Speg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108802.1 | NP_001102272.1 | 11 | 3442 | ||
| XM_006245252.2 | XP_006245314.1 | 13 | 3404 | |||
| XM_006245255.2 | XP_006245317.1 | 6 | 3200 | |||
| XM_006245256.2 | XP_006245318.1 | 6 | 1229 | |||
| XM_008767267.1 | XP_008765489.1 | 11 | 3504 | |||
| XM_008767268.1 | XP_008765490.1 | 11 | 3281 | |||
| XM_008767269.1 | XP_008765491.1 | 12 | 3898 | |||

Speg
Genomic Map