siRNA Details
| Gene Symbol: Col4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135759.1 | NP_001129231.1 | 25 | 1597 | ||
| XM_006245259.2 | XP_006245321.1 | 25 | 1820 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col4a3| Gene Symbol: Col4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135759.1 | NP_001129231.1 | 25 | 1597 | ||
| XM_006245259.2 | XP_006245321.1 | 25 | 1820 | |||