siRNA Details
| Gene Symbol: Agfg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135596.1 | NP_001129068.1 | 5 | 908 | ||
| GenBank mRNA | BC099202.1 | - | 5 | 908 | 7384071 | |
| FQ211754.1 | - | 6 | 751 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agfg1| Gene Symbol: Agfg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135596.1 | NP_001129068.1 | 5 | 908 | ||
| GenBank mRNA | BC099202.1 | - | 5 | 908 | 7384071 | |
| FQ211754.1 | - | 6 | 751 | |||