siRNA Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025025.1 | NP_001020196.1 | 10 | 974 | ||
| GenBank mRNA | BC079402.1 | - | 10 | 957 | 7112774 | |
| BC091226.1 | - | 10 | 974 | 7131918 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Daw1| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025025.1 | NP_001020196.1 | 10 | 974 | ||
| GenBank mRNA | BC079402.1 | - | 10 | 957 | 7112774 | |
| BC091226.1 | - | 10 | 974 | 7131918 | ||