siRNA Details
| Gene Symbol: Thap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005564.1 | NP_001005564.1 | 2 | 592 | ||
| GenBank mRNA | BC081882.1 | - | 2 | 592 | 7108379 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap4| Gene Symbol: Thap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005564.1 | NP_001005564.1 | 2 | 592 | ||
| GenBank mRNA | BC081882.1 | - | 2 | 592 | 7108379 | |