siRNA Details
| Gene Symbol: Ing5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108810.1 | NP_001102280.1 | 2 | 192 | ||
| GenBank mRNA | BC167048.1 | - | 2 | 63 | 9100546 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ing5| Gene Symbol: Ing5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108810.1 | NP_001102280.1 | 2 | 192 | ||
| GenBank mRNA | BC167048.1 | - | 2 | 63 | 9100546 | |