siRNA Details
| Gene Symbol: Tbcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271364.1 | NP_001258293.1 | 28 | 2449 | ||
| XM_006247861.2 | XP_006247923.1 | 28 | 2510 | |||
| XM_006247864.2 | XP_006247926.1 | 28 | 2510 | |||
| XM_008768500.1 | XP_008766722.1 | 26 | 2426 | |||
| XM_008768501.1 | XP_008766723.1 | 29 | 2512 | |||
| XM_008768502.1 | XP_008766724.1 | 24 | 1917 | |||
| GenBank mRNA | FQ213737.1 | - | 4 | 301 | ||

Tbcd
Genomic Map