siRNA Details
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135893.1 | NP_001129365.1 | 41 | 7433 | ||
NM_001135923.1 | NP_001129395.1 | 40 | 7306 | |||
XM_006256666.2 | XP_006256728.1 | 43 | 7705 | |||
XM_006256667.2 | XP_006256729.1 | 44 | 7713 | |||
XM_006256668.2 | XP_006256730.1 | 43 | 7698 | |||
XM_006256669.2 | XP_006256731.1 | 40 | 7344 | |||
XM_006256670.2 | XP_006256732.1 | 43 | 7949 | |||
XM_006256671.2 | XP_006256733.1 | 40 | 7330 | |||
XM_008773061.1 | XP_008771283.1 | 43 | 7964 |
