siRNA Details
| Gene Symbol: Otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037496.1 | NP_001032585.1 | 6 | 984 | ||
| XM_006256737.2 | XP_006256799.1 | 6 | 1634 | |||
| XM_006256738.2 | XP_006256800.1 | 6 | 1633 | |||
| XM_006256739.2 | XP_006256801.1 | 6 | 1632 | |||
| XM_006256740.2 | XP_006256802.1 | 7 | 1504 | |||
| XM_006256741.2 | XP_006256803.1 | 7 | 1503 | |||
| GenBank mRNA | BC110054.1 | - | 6 | 984 | 7446041 | |

Otud5
Genomic Map