siRNA Details
| Gene Symbol: Chrdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199502.1 | NP_955796.1 | 8 | 1107 | ||
| XM_006257373.2 | XP_006257435.1 | 8 | 1134 | |||
| GenBank mRNA | AB080636.1 | - | 8 | 1107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrdl1| Gene Symbol: Chrdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199502.1 | NP_955796.1 | 8 | 1107 | ||
| XM_006257373.2 | XP_006257435.1 | 8 | 1134 | |||
| GenBank mRNA | AB080636.1 | - | 8 | 1107 | ||