siRNA Details
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037546.1 | NP_001032635.1 | 1664 | |||
| XM_008773111.1 | XP_008771333.1 | 2268 | ||||
| GenBank mRNA | BC110051.1 | - | 1664 | 7459258 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008001, BC011462, AK148151, U22325, BC004838, and others.

Fgd1
Genomic Map