siRNA Details
| Gene Symbol: Gemin8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007756.1 | NP_001007757.1 | 4 | 797 | ||
| GenBank mRNA | BC079160.1 | - | 4 | 797 | 7129380 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gemin8| Gene Symbol: Gemin8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007756.1 | NP_001007757.1 | 4 | 797 | ||
| GenBank mRNA | BC079160.1 | - | 4 | 797 | 7129380 | |