siRNA Details
| Gene Symbol: Pir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009474.1 | NP_001009474.1 | 5, 6 | 616 | ||
| GenBank mRNA | BC088290.1 | - | 5, 6 | 616 | 7322321 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pir| Gene Symbol: Pir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009474.1 | NP_001009474.1 | 5, 6 | 616 | ||
| GenBank mRNA | BC088290.1 | - | 5, 6 | 616 | 7322321 | |