siRNA Details
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009474.1 | NP_001009474.1 | 10 | 930 | ||
GenBank mRNA | BC088290.1 | - | 10 | 930 | 7322321 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009474.1 | NP_001009474.1 | 10 | 930 | ||
GenBank mRNA | BC088290.1 | - | 10 | 930 | 7322321 |