siRNA Details
| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135877.1 | NP_001129349.1 | 7, 8 | 844 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf7l| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135877.1 | NP_001129349.1 | 7, 8 | 844 | ||