siRNA Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FM134560.1 | - | 3 | 321 | ||
| FM135509.1 | - | 4 | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gla| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FM134560.1 | - | 3 | 321 | ||
| FM135509.1 | - | 4 | 450 | |||