siRNA Details
| Gene Symbol: Polr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_343922.6 | XP_343923.3 | 9 | 1832 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr2a| Gene Symbol: Polr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_343922.6 | XP_343923.3 | 9 | 1832 | ||