siRNA Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105883.1 | NP_001099353.1 | 6 | 1179 | ||
| XM_006248338.2 | XP_006248400.1 | 6 | 1217 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl3| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105883.1 | NP_001099353.1 | 6 | 1179 | ||
| XM_006248338.2 | XP_006248400.1 | 6 | 1217 | |||