siRNA Details
| Gene Symbol: Fbxo40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248404.2 | XP_006248466.1 | 4 | 1370 | ||
| XM_344023.7 | XP_344024.5 | 3 | 1255 | |||
| GenBank mRNA | FQ217730.1 | - | 1395 | |||
| FQ223618.1 | - | 1395 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo40| Gene Symbol: Fbxo40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248404.2 | XP_006248466.1 | 4 | 1370 | ||
| XM_344023.7 | XP_344024.5 | 3 | 1255 | |||
| GenBank mRNA | FQ217730.1 | - | 1395 | |||
| FQ223618.1 | - | 1395 | ||||