siRNA Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108848.1 | NP_001102318.1 | 5 | 605 | ||
| GenBank mRNA | AB212679.1 | - | 4 | 453 | ||
| BC168972.1 | - | 5 | 611 | 9034405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elavl1| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108848.1 | NP_001102318.1 | 5 | 605 | ||
| GenBank mRNA | AB212679.1 | - | 4 | 453 | ||
| BC168972.1 | - | 5 | 611 | 9034405 | ||