siRNA Details
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012475.1 | NP_001012493.1 | 17 | 1788 | ||
GenBank mRNA | AY906861.1 | - | 17 | 1788 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012475.1 | NP_001012493.1 | 17 | 1788 | ||
GenBank mRNA | AY906861.1 | - | 17 | 1788 |